missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pentaerythritol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 129870050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pentaerythritol | |
257.0°C to 264.0°C | |
276.0°C (30.0 mmHg) | |
Authentic | |
Plastic Drum | |
C(CH2OH)4 | |
01,528 | |
pentaerythritol, pentek, 2,2-bis hydroxymethyl propane-1,3-diol, tetramethylolmethane, monopentaerythritol, auxinutril, maxinutril, monopentek, pentaerythrite, penetek | |
WXZMFSXDPGVJKK-UHFFFAOYSA-N | |
2,2-bis(hydroxymethyl)propane-1,3-diol | |
8285 | |
98% |
115-77-5 | |
White | |
240°C | |
97.5% min. (GC) | |
C5H12O4 | |
5 kg | |
15,7221 | |
Solubility in water: 1 g/18 mL water (15°C). Other solubilities: slightly soluble in alcohol and formamide, insoluble in benzene, carbon tetrachloride, ether,, acetone and petroleum ether | |
C(C(CO)(CO)CO)O | |
136.15 | |
136.15 | |
Crystals |
Chemical Identifiers
115-77-5 | |
136.15 | |
pentaerythritol, pentek, 2,2-bis hydroxymethyl propane-1,3-diol, tetramethylolmethane, monopentaerythritol, auxinutril, maxinutril, monopentek, pentaerythrite, penetek | |
2,2-bis(hydroxymethyl)propane-1,3-diol |
C5H12O4 | |
WXZMFSXDPGVJKK-UHFFFAOYSA-N | |
8285 | |
C(C(CO)(CO)CO)O |
Safety and Handling
EINECSNumber : 204-104-9
RUO – Research Use Only