missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pentaerythritol, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A105200B
Description
Pentaerythritol is used as chemical intermediate for alkylated resins, synth lubricants, pentaerythritol tetranitrate & resin & tall oil esters. It is also used in coating, dynamite and plastic industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPentaerythritol is used as chemical intermediate for alkylated resins, synth lubricants, pentaerythritol tetranitrate & resin & tall oil esters. It is also used in coating, dynamite and plastic industry.
Solubility
Soluble in Water: 0.1 g/mL
Notes
Hygroscopic. Incompatible with strong acids, strong oxidizing agents, acid chlorides, acid anhydrides.
Specifications
Pentaerythritol | |
260°C to 264°C | |
276°C (30 mmHg) | |
C5H12O4 | |
1000 g | |
Hygroscopic | |
pentaerythritol, pentek, 2,2-bis hydroxymethyl propane-1,3-diol, tetramethylolmethane, monopentaerythritol, auxinutril, maxinutril, monopentek, pentaerythrite, penetek | |
WXZMFSXDPGVJKK-UHFFFAOYSA-N | |
2,2-bis(hydroxymethyl)propane-1,3-diol | |
8285 | |
≥98% |
115-77-5 | |
1.396 | |
240°C (464°F) | |
MFCD00004692 | |
1679274 | |
14,7111 | |
Soluble in Water: 0.1g/mL | |
C(C(CO)(CO)CO)O | |
136.147 | |
136.15 |
Chemical Identifiers
115-77-5 | |
136.147 | |
WXZMFSXDPGVJKK-UHFFFAOYSA-N | |
8285 | |
C(C(CO)(CO)CO)O |
C5H12O4 | |
MFCD00004692 | |
pentaerythritol, pentek, 2,2-bis hydroxymethyl propane-1,3-diol, tetramethylolmethane, monopentaerythritol, auxinutril, maxinutril, monopentek, pentaerythrite, penetek | |
2,2-bis(hydroxymethyl)propane-1,3-diol |
Safety and Handling
H500
EINECSNumber : 204-104-9
RTECSNumber : RZ2490000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only