missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentadecyl Acetate 95.0+%, TCI America™

Supplier:  TCI America P17005G

 View more versions of this product

Catalog No. P17005G



Description

Description

Specifications

Specifications

Pentadecyl Acetate
15°C
230°C
MFCD00056204
Acetic Acid Pentadecyl Ester
CCCCCCCCCCCCCCCOC(=O)C
270.457
270.46
Liquid
629-58-3
Colorless
C17H34O2
5 g
AAAIZLQILBFRTJ-UHFFFAOYSA-N
pentadecyl acetate
522077
≥95.0% (GC)

Chemical Identifiers

629-58-3
270.457
AAAIZLQILBFRTJ-UHFFFAOYSA-N
522077
CCCCCCCCCCCCCCCOC(=O)C
C17H34O2
MFCD00056204
Acetic Acid Pentadecyl Ester
pentadecyl acetate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0735

TSCA : No

SDS
Documents

Documents

Promotions

Promotions