missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pentadecanolide, 98%

Manufacturer:  Alfa Aesar™ A1134114

 View more versions of this product

Catalog No. AAA1134114


 

Description

Description

Applications
Pentadecanolide is a monomer used to make PGA-co-pentadecalactone polymers to be used in the development of directly compressed prolonged release tablets of slightly soluble drugs such as ketoprofen. Pentadecanolide may be used for the synthesis of poly(pentadecanolide) (PPDL) by Novozyme 435 catalyzed ring-opening polymerization. It may be employed as starting reagent for the synthesis of terminally-branched iso-fatty acids (iso-C15-C17).

Solubility
Soluble in alcohol, dipropylene glycol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

Pentadecanolide
98%
MFCD00039667
14,3910
FKUPPRZPSYCDRS-UHFFFAOYSA-N
oxacyclohexadecan-2-one
235414
33°C to 37°C
25g
106-02-5
C15H28O2
143710
Cyclopentadecanolide; 15-Hydroxypentadecanoic acid lactone
C1CCCCCCCOC(=O)CCCCCC1
240.387
240.39
137°C (2mmHg)
>110°C (230°F)

Chemical Identifiers

106-02-5
240.387
FKUPPRZPSYCDRS-UHFFFAOYSA-N
235414
C15H28O2
MFCD00039667
Cyclopentadecanolide; 15-Hydroxypentadecanoic acid lactone
Safety and Handling

Safety and Handling

RTECSNumber : RN9775000

TSCA : TSCA

SDS Documents