missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentadecafluorooctanenitrile, TCI America™

Manufacturer:  TCI America P24835G

 View more versions of this product

Catalog No. P24835G



Specifications

Specifications

Pentadecafluorooctanenitrile
≥96.0% (GC)
MFCD00039481
FULLNJNBFUCVES-UHFFFAOYSA-N
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanenitrile
69536
Clear Liquid at 20°C
102°C
647-12-1
C8F15N
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluorooctanenitrile, Perfluorooctanonitrile
C(#N)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
395.071
395.07
Yellow
5g

Chemical Identifiers

647-12-1
395.071
FULLNJNBFUCVES-UHFFFAOYSA-N
69536
C(#N)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
C8F15N
MFCD00039481
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluorooctanenitrile, Perfluorooctanonitrile
2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanenitrile
Safety and Handling

Safety and Handling

TSCA : No

Documents