missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentadecafluoroheptyl Iodide (stabilized with Na2S2O3), TCI America™

Manufacturer:  TCI America P183925G

 View more versions of this product

Catalog No. P183925G



Specifications

Specifications

Pentadecafluoroheptyl Iodide (stabilized with Na2S2O3)
Yellow
25g
C7F15I
Perfluoroheptyl Iodide
C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(C(F)(F)I)(F)F)(F)F)(F)F
495.958
495.96
≥98.0% (GC)
138°C
-8°C
335-58-0
MFCD00013712
AHUMDLIBMIYQMU-UHFFFAOYSA-N
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7-pentadecafluoro-7-iodoheptane
67633
Clear Liquid at 20°C

Chemical Identifiers

335-58-0
495.958
AHUMDLIBMIYQMU-UHFFFAOYSA-N
67633
C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(C(F)(F)I)(F)F)(F)F)(F)F
C7F15I
MFCD00013712
Perfluoroheptyl Iodide
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7-pentadecafluoro-7-iodoheptane
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0090

TSCA : Yes

Documents