missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentachloropyridine 98%, ACROS Organics™

Manufacturer:  Acros Organics 160220250

 View more versions of this product

Catalog No. AC160220250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Pentachloropyridine
Glass bottle
124.0°C to 126.0°C
98%
98%
(C5N)Cl5
20, 232
DNDPLEAVNVOOQZ-UHFFFAOYSA-N
2,3,4,5,6-pentachloropyridine
16584
Flakes
Authentic
White
25g
2176-62-7
C5Cl5N
MFCD00006230
pentachloropyridine, perchloropyridine, pyridine, pentachloro, pyridine, 2,3,4,5,6-pentachloro, unii-6mm852ka6t, 2,3,4,5,6-pentachlorpyridine, dsstox_cid_2179, dsstox_rid_76515, dsstox_gsid_22179, pentachlorpyridine
C1(=C(C(=NC(=C1Cl)Cl)Cl)Cl)Cl
251.312
251.33
98%

Chemical Identifiers

2176-62-7
251.312
DNDPLEAVNVOOQZ-UHFFFAOYSA-N
16584
C1(=C(C(=NC(=C1Cl)Cl)Cl)Cl)Cl
C5Cl5N
MFCD00006230
pentachloropyridine, perchloropyridine, pyridine, pentachloro, pyridine, 2,3,4,5,6-pentachloro, unii-6mm852ka6t, 2,3,4,5,6-pentachlorpyridine, dsstox_cid_2179, dsstox_rid_76515, dsstox_gsid_22179, pentachlorpyridine
2,3,4,5,6-pentachloropyridine
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 218-535-5

RTECSNumber : UT7000000

TSCA : TSCA

SDS Documents