Please login to your online account to display your discounted pricing

Pentachloropyridine 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Pentachloropyridine
Name Note 98%
CAS 2176-62-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5Cl5N
Linear Formula (C5N)Cl5
Formula Weight 251.33
MDL Number MFCD00006230
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC160220250

  SDS
fisher scientific acros
160220250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC160220050

  SDS
thermo fisher scientific
160220050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC160221000

  SDS
fisher scientific acros
160221000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Flakes
Assay ≥97.5%
Formula Weight 251.33
Quantity 25g
Color White to off-white
Beilstein 20, 232
EINECS Number 218-535-5
TSCA TSCA
Melting Point 124° to 126°C
RTECS Number UT7000000
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Pentachloropyridine
Name Note 98%
Assay Percent Range 98%
Molecular Formula C5Cl5N
Linear Formula (C5N)Cl5
MDL Number MFCD00006230
CAS 2176-62-7
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.