Please login to your online account to display your discounted pricing

Alfa Aesar™ Pentachloroethane, 96%

Chemical Identifiers

Chemical Name or Material Pentachloroethane
CAS 76-01-7
Molecular Formula C2HCl5
Linear Formula CCl3CHCl2
Formula Weight 202.3
MDL Number MFCD00000846
UN Number UN1669
Beilstein 1736845
Merck Index 14,7108
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4447509

  SDS
alfa aesar™
4447509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4447518

  SDS
alfa aesar™
4447518
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4447530

  SDS
alfa aesar™
4447530
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 202.3
Chemical Name or Material Pentachloroethane
Density 1.68g/mL
TSCA TSCA
Physical Form Liquid
Assay 96%
Molecular Formula C2HCl5
Linear Formula CCl3CHCl2
MDL Number MFCD00000846
CAS 76-01-7
UN Number UN1669
Beilstein 1736845
Merck Index 14,7108
Refractive Index 1.5030
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H372-H351
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Color Colorless
Melting Point −29°C
Boiling Point 162°C
Quantity 10g
Odor Like chlorine
Health Hazard 3 GHS P Statement
P260-P281-P264-P308+P313-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
Wash thoroughly after handling.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PENTACHLOROETHANE
RTECS Number KI6300000
Safety and Handling

Danger

GHS P Statement
P260-P281-P264-P308+P313-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
Wash thoroughly after handling.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H372-H351
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.