missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Penta-O-acetyl-beta-D-glucopyranose, TCI America™

Manufacturer:  TCI America P0028100G

 View more versions of this product

Catalog No. P0028100G



Specifications

Specifications

Penta-O-acetyl-beta-D-glucopyranose
132°C
604-69-3
MFCD00006597
LPTITAGPBXDDGR-IBEHDNSVSA-N
[(2R,3R,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
2724702
Crystal-Powder at 20°C
White
100g
C16H22O11
beta-d-glucose pentaacetate, penta-o-acetyl-beta-d-glucopyranose, unii-v833p95coc, 1,2,3,4,6-penta-o-acetyl-beta-d-glucopyranose, 1,2,3,4,6-penta-o-acetyl-b-d-glucopyranose, beta-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, 2s,3r,4s,5r,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, glucopyranose, pentaacetate, .beta.-d, penta-o-acetyl-&beta
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
390.341
390.34
≥99.0% (GC)

Chemical Identifiers

604-69-3
390.341
LPTITAGPBXDDGR-IBEHDNSVSA-N
2724702
CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
C16H22O11
MFCD00006597
beta-d-glucose pentaacetate, penta-o-acetyl-beta-d-glucopyranose, unii-v833p95coc, 1,2,3,4,6-penta-o-acetyl-beta-d-glucopyranose, 1,2,3,4,6-penta-o-acetyl-b-d-glucopyranose, beta-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, 2s,3r,4s,5r,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, glucopyranose, pentaacetate, .beta.-d, penta-o-acetyl-&beta
[(2R,3R,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
Safety and Handling

Safety and Handling

EINECSNumber : (8)-0053

TSCA : No

Documents