Penicillin-Streptomycin, Dual Antibiotic Solution, MP Biomedicals
Dual antibiotic solution Concentration: Penicillin- 5,000 IU/mL, Streptomycin- 5mg/mL Effective against gram-positive and gram-negative bacteria
Manufacturer:  Mp Biomedicals 1670049
Catalog No. ICN1670049
Specifications
Specifications
Penicillin-Streptomycin
Pen-Strep
100mL
Documents