Please login to your online account to display your discounted pricing

Penicillin G sodium salt, 99.7%,USP, MP Biomedicals™


Quantity

Manufacturer: mp biomedicals  10054801

 View more versions of this product

Catalog No. ICN10054801

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 356.4
Color White
Quantity
Purity Grade USP
Chemical Name or Material Penicillin G Sodium Salt
Assay Percent Range 0.997
Recommended Storage Store at 2° to 8°C.
Molecular Formula C16H17N2O4SNa
CAS 69-57-8
Synonym Benzylpenicillin
Storage Note 1 Protect from light.

Specifications

Chemical Name or Material Penicillin G Sodium Salt
CAS 69-57-8
Purity Grade USP
Physical Form Powder
Assay Percent Range 0.997
Molecular Formula C16H17N2O4SNa
Formula Weight 356.4
Synonym Benzylpenicillin
Accessories

Store at 2° to 8°C.

Protect from light.