Please login to your online account to display your discounted pricing

Penicillin G sodium salt, 99.7%,USP, MP Biomedicals™

$22.67 - $22.67

Chemical Identifiers

Chemical Name or Material Penicillin G Sodium Salt
CAS 69-57-8
Purity Grade USP
Physical Form Powder
Assay Percent Range 0.997
Molecular Formula C16H17N2O4SNa
Formula Weight 356.4
Synonym Benzylpenicillin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10054801

 
mp biomedicals
10054801
Each for $22.67
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 356.4
Color White
Quantity
Purity Grade USP
Recommended Storage Store at 2° to 8°C.
Assay Percent Range 0.997
Chemical Name or Material Penicillin G Sodium Salt
CAS 69-57-8
Molecular Formula C16H17N2O4SNa
Synonym Benzylpenicillin
Storage Note 1 Protect from light.
Safety and Handling

Store at 2° to 8°C.

Protect from light.