Learn More
Penicillin G sodium salt, Thermo Scientific Chemicals
Penicillin G sodium salt, CAS # 69-57-8, also called benzylpenicillin, is a β-lactam antibiotic active against gram-positive and gram-negative bacteria.
Supplier: Thermo Scientific Chemicals J6303206
Description
Penicillin G sodium salt is used as a β-lactam antibiotic. Penicillin G can be used as a selective agent in several types of isolation media. It has been used to study the diagnostic and therapeutic implications of gentamicin-resistant Enterococcus faecalis sequence type 6 with reduced penicillin susceptibility and in cell culture both alone and combined with streptomycin and other antibiotics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Penicillin G Sodium Salt | |
Powder | |
C16H17N2NaO4S | |
3834217 | |
Benzylpenicillin sodium salt | |
FCPVYOBCFFNJFS-LQDWTQKMSA-M | |
sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate | |
23668834 | |
356.37 | |
5 g |
Penicillin G sodium salt | |
69-57-8 | |
MFCD00069666 | |
14,7094 | |
Soluble in water (100mg/ml),alcohol,glycerol,DMSO (71mg/ml at 25°C),and ethanol (< 1mg/ml at 25°C). | |
CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+] | |
356.372 | |
CHEBI:51765 | |
White |
Chemical Identifiers
69-57-8 | |
356.372 | |
FCPVYOBCFFNJFS-LQDWTQKMSA-M | |
23668834 | |
sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
C16H17N2NaO4S | |
MFCD00069666 | |
Benzylpenicillin sodium salt | |
CHEBI:51765 | |
CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+] |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
EINECSNumber : 200-710-2
RTECSNumber : XH9800000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only