missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Penicillin G potassium salt, 100 mg/mL in distilled water, sterile-filtered, Alfa Aesar™

$84.70 - $120.86

Chemical Identifiers

CAS 113-98-4
Molecular Formula C16H17KN2O4S
Molecular Weight (g/mol) 372.48
MDL Number MFCD00036193
InChI Key IYNDLOXRXUOGIU-LQDWTQKMSA-M
PubChem CID 23664709
ChEBI CHEBI:7963
IUPAC Name potassium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67333AD
View Documents
Thermo Fisher Scientific
J67333AD
50mL
Each for $84.70
 
AAJ67333AE
View Documents
Thermo Fisher Scientific
J67333AE
100mL
Each for $120.86
 
Description

Description

Specifications

Chemical Identifiers

113-98-4
372.48
IYNDLOXRXUOGIU-LQDWTQKMSA-M
CHEBI:7963
CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[K+]
C16H17KN2O4S
MFCD00036193
23664709
potassium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate

Specifications

113-98-4
MFCD00036193
CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[K+]
372.48
CHEBI:7963
100mg/mL in distilled water
Penicillin G potassium salt, sterile-filtered
C16H17KN2O4S
IYNDLOXRXUOGIU-LQDWTQKMSA-M
potassium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
23664709
Liquid
50mL
Safety and Handling

Safety and Handling

P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c

H317-H334

EINECSNumber : 204-038-0

RTECSNumber : XH9700000

TSCA : Yes

Recommended Storage : Store at -20°C

SDS