missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pendimethalin 98.0+%, TCI America™
Supplier: TCI America P196425G
Specifications
Pendimethalin | |
58°C | |
MFCD00055332 | |
3077 | |
CHIFOSRWCNZCFN-UHFFFAOYSA-N | |
3,4-dimethyl-2,6-dinitro-N-pentan-3-ylaniline | |
38479 | |
281.31 | |
Crystalline Powder |
40487-42-1 | |
C13H19N3O4 | |
25 g | |
pendimethalin, pendimethaline, penoxaline, prowl, herbadox, penoxalin, phenoxalin, accotab, stomp, way up | |
CCC(CC)NC1=C(C=C(C(=C1[N+](=O)[O-])C)C)[N+](=O)[O-] | |
281.312 | |
CHEBI:83569 | |
≥98.0% (GC) |
Chemical Identifiers
40487-42-1 | |
281.312 | |
CHIFOSRWCNZCFN-UHFFFAOYSA-N | |
38479 | |
3,4-dimethyl-2,6-dinitro-N-pentan-3-ylaniline |
C13H19N3O4 | |
MFCD00055332 | |
pendimethalin, pendimethaline, penoxaline, prowl, herbadox, penoxalin, phenoxalin, accotab, stomp, way up | |
CHEBI:83569 | |
CCC(CC)NC1=C(C=C(C(=C1[N+](=O)[O-])C)C)[N+](=O)[O-] |
Safety and Handling
RTECSNumber : BX5470000
TSCA : No