missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ PELTOR™ Earmuffs, Green

3M™ PELTOR™ Earmuffs X1 are our lightweight hearing protectors with comfortable, soft ear cushions.

Manufacturer:  3M™ 7000104070

Catalog No. 17-000-631




Description

Description

  • Reduces noise levels up to 27 dB
  • Wide flat headband is electrically insulted making the earmuffs electrically resistant or 'dielectric'
  • Colour-coding makes it easy to identify the level of protection being worn
  • Slim cups offer improved compatibility with other safety equipment
  • Wide soft ear cushions help workers stay comfortable
  • Twin headband design provides heat dispersion and internal stainless steel design offers consistent pressure and comfortable fit over an 8 hour period
  • Designed to optimise weight distribution and balance for outstanding comfort
  • ABS plastic cups are rigid, high strength and impact resistant
  • Thermoplastic polyurethane (TPU) soft outer cup coating protects from abrasion and bumps

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Specifications

Specifications

Black, Green
Foam
Black
Composite Finishing, Final Finish, Grinding, Machine Operations, Paint Preparation, Painting, Welding
1 Ea.
X1 Series
Yes
Over-the-head
Yes
Class A
Wire
X1A
22 dB
Black
ABS/TPU, PU Foam and PVC
SDS
Documents
Product Certifications
Promotions

Promotions