Please login to your online account to display your discounted pricing

Pelican™ StealthLite™ Flashlight and Rechargeable System

IPX7 rating

$54.93 - $54.93

Specifications

Battery Alkaline
Certifications/Compliance CSA, FM, Class I, Division 2, ETL, ANSI/NEMA FL 1
Product Type Flashlight
Weight (Metric) 0.2kg
Includes 4 AA alkaline batteries
View More Specs
Includes:  Battery-Powered System: Four “AA” alkaline batteries.
Rechargeable System: StealthLite flashlight, desk/dash mount base, 110V charger, and high-capacity NiMH battery pack.
Products
Catalog Number Mfr. No. Battery Run Time For Use With (Application) Length (Metric) Color Price Quantity    

19-086789

 
pelican™
2400010150
3.45 hr. The tough resin body is corrosion proof and resistant to extreme temperatures, xenon laser spot lamp module with white collimated beam one-hand safety switch and double shock mounted battery compartment 16.48cm Orange Each for $54.93
Description & Specifications

Specifications

Battery Alkaline
Certifications/Compliance CSA, FM, Class I, Division 2, ETL, ANSI/NEMA FL 1
Product Type Flashlight
Weight (Metric) 0.2kg
Includes 4 AA alkaline batteries
Light Source Xenon
Voltage 6V

  • 3.75 hours burn time
  • Lexan EXL resin
  • Black lanyard

Battery-Powered System

  • Xenon Laser Spot lamp module with white collimated beam One-hand safety switch and double-shock-mounted battery compartment
Rechargeable System
    High-performance Xenon Laser Spot lamp module has 50% higher candlepower than module for battery-powered StealthLite

Length: 6.5 in. (16.5cm). Weight (with batteries): 7.1 oz. (201g).

CSA and FM approved for certain Class I, Division 2 hazardous locations; ETL marked.