Please login to your online account to display your discounted pricing

Pelican™ L4™ 1830 LED Lights

Lightweight, slim-profile design fits in pocket

$26.16 - $26.45

Specifications

Battery Alkaline
Certifications/Compliance FM, Class I, Division 1 hazardous locations, CE
Product Type 1830 LED Light
Weight (English) 1.2 oz.
Weight (Metric) 0.03kg
View More Specs
Includes:  Three “AAAA” batteries
Products
Catalog Number Mfr. No. Color Price Quantity    

19-150-0096

 
pelican™
1830010110
Black Each for $26.45
Pack of 12 Each for $282.60

19-150-0097

 
pelican™
1830010245
Yellow Each for $26.16
Pack of 12 Each for $279.48
Description & Specifications

Specifications

Battery Alkaline
Certifications/Compliance FM, Class I, Division 1 hazardous locations, CE
Product Type 1830 LED Light
Weight (English) 1.2 oz.
Weight (Metric) 0.03kg
For Use With (Application) Lightweight, slim-profile design fits in pocket
Includes Three AAAA batteries
Lamp Life 10000 hr.
Length (English) 6.2 in.
Length (Metric) 15.7cm
Lumen 5lm
Material Xenoy™ Polycarbonate Resin
Voltage 4.5V

  • 32-hour burn time
  • Handy and unbelievably bright
  • Tail-button switch provides quick ON/OFF or momentary activation
  • Clip molded to Xenoy™ body
  • Molded of polycarbonate resin—safe to use in electrical environments

Length: 6.2 in. (15.7cm). Weight (with batteries): 1.2 oz. (34g).

FM listed for certain Class I, Division 1 hazardous locations; CE marked.