Please login to your online account to display your discounted pricing

Peel-Off™ China Markers


Great for writing on china, glass, metal or plastic.

Manufacturer: fisher science education  852-059

 View more versions of this product

Catalog No. S01693

  • / Dozen of 12 Each
 

Description & Specifications

Specifications

Product Type Sharpie™ Peel-Off™ China Marker
Color Yellow
For Use With (Application) Great for writing on china, glass, metal or plastic
Material (Internal) Wax
Quantity 12
Tip Style Medium