Peel-Off™ China Markers
Great for writing on china, glass, metal or plastic.
$41.00 - $41.00
Specifications
For Use With (Application) | Great for writing on china, glass, metal or plastic |
---|---|
Material (Internal) | Wax |
Product Type | Sharpie™ Peel-Off™ China Marker |
Quantity | 12 |
Tip Style | Medium |
Description
- No sharpening required — just peel the tear-string to reveal more marker
- Cleanup is simple; marks are easily erased with a damp cloth
- Fade- and water-resistant
- Certified AP nontoxic
- Medium or conical point
- Ink type: wax, nonretractable, nonwashable
Order Info
Sold as a dozen.
Specifications
Great for writing on china, glass, metal or plastic | |
Sharpie™ Peel-Off™ China Marker | |
Medium |
Wax | |
12 |