missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Pectin Citrus

$37.16 - $147.42

Chemical Identifiers

CAS 9000-69-5
MDL Number MFCD00081838
Synonym Poly-D-galacturonic acid methyl ester
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6102122 View Documents Alfa Aesar™
J6102122
100g Each for $37.16
 
AAJ6102136 View Documents Alfa Aesar™
J6102136
500g Each for $147.42
 
Description

Description

Applications
Pectin Citrus is promoted in dietary supplement form as an alternative cancer treatment. It can also been used to treat prostate cancer and melanoma. It can also be used as a gelling agent, filler, and stabilizer. It Supports celluar health, promotes immune function, supports cardiovascular health, supports healthy function of multiple organs, Safely chelates heavy metals and toxins.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Chemical Identifiers

9000-69-5
MFCD00081838
Poly-D-galacturonic acid methyl ester

Specifications

Brown to Yellow
9000-69-5
14,7063
Powder
100g
MFCD00081838
Poly-D-galacturonic acid methyl ester
Pectin Citrus
Safety and Handling

Safety and Handling

RTECSNumber : RX4280000

TSCA : TSCA