Invitrogen™ PDK4 (PDKH4) Recombinant Human Protein DFS Item
Inhhibition of PDHKs is viewed as a potential therapeutic treament for type II diabetes.
Manufacturer:  Invitrogen™ LSPV5711
 View more versions of this product
Catalog No. PV5711
Description
Description
PDK4 (PDHK4), along with the other PDHKs, regulates glucose metabolism by phosphorylating the E1 alpha subunit of the mitochondrial pyruvate dehydrogenase complex.
Specifications
Specifications
NP_002603.1
Kinase Assay
Frozen
73.9kDa
Kinase proteins are shipped on dry ice. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
PDHK Kinase Family
Yes
Baculovirus
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol.
1mg
RUO - research use only
PDK-4 (PDKH4)
Human
GST Tag