missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ PDK4 (PDKH4) Recombinant Human Protein DFS Item

Inhhibition of PDHKs is viewed as a potential therapeutic treament for type II diabetes.

$1,976.00 - $8,390.00

Specifications

Format Frozen
Host Species Human
Protein Tag GST Tag
Recombinant Yes
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PR8253A View Documents Invitrogen™
LSPR8253A
100μg Each for $1,976.00
 
PV5711 View Documents Invitrogen™
LSPV5711
1mg Each for $8,390.00
 
Description

Description

PDK4 (PDHK4), along with the other PDHKs, regulates glucose metabolism by phosphorylating the E1 alpha subunit of the mitochondrial pyruvate dehydrogenase complex.

Specifications

Specifications

Frozen
Human
GST Tag
Yes