missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ PDGFRA [V561D] Recombinant Human Protein
PDGFRA is a cell surface receptor tyrosine kinase.
$2650.00 - $11260.00
Specifications
Accession Number | NP_006197, V561D |
---|---|
Formulation | 50mM Tris, pH 7.5, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT, 50 % Glycerol |
Molecular Weight (g/mol) | 89.8kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PR7732A
|
Invitrogen™
PR7732A |
100μg |
Each for $2,650.00
|
|
|||||
PV4681
|
Invitrogen™
PV4681 |
1mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
PDGFRA binds members of the platelet-derived growth factor family. The PDGFRA V561D mutation is observed in gastrointestinal stromal tumors (GIST).
Specifications
NP_006197, V561D | |
89.8kDa | |
RUO - research use only | |
PDGFRA V561D, PDGFR alpha | |
Human | |
GST Tag | |
Frozen |
50mM Tris, pH 7.5, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT, 50 % Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
PDGFR Kinase Family | |
Recombinant | |
Baculovirus |