PDGFRA [V561D] Recombinant Human Protein, Invitrogen™ DFS Item
PDGFRA is a cell surface receptor tyrosine kinase.
Manufacturer:  Life Technologies LSPV4681
 View more versions of this product
Catalog No. PV4681
Description
Description
PDGFRA binds members of the platelet-derived growth factor family. The PDGFRA V561D mutation is observed in gastrointestinal stromal tumors (GIST).
Specifications
Specifications
NP_006197, V561D
Frozen
89.8kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
PDGFR Kinase Family
Yes
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
50mM Tris, pH 7.5, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT, 50 % Glycerol
1mg
RUO - research use only
PDGFRA V561D, PDGFR alpha
Baculovirus
Human
GST Tag
Documents