missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PDGFRA [T674I] Recombinant Human Protein, Invitrogen™ DFS Item

PDGFRA T674I is a Gleevec resistant form of PDGFRA.

Manufacturer:  Life Technologies LSPV3847

 View more versions of this product

Catalog No. PV3847



Description

Description

PDGFRA is a cell surface receptor tyrosine kinase which binds members of the platelet-derived growth factor family.

Specifications

Specifications

NP_006197, T674T
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
89.8kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Recombinant, Cytoplasmic
PDGFRA T674I, PDGFR alpha
Kinase Assay
Baculovirus
aa 550-1089; T674I
Yes
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
5156
10μg
RUO - research use only
Receptor Tyrosine Kinases
platelet-derived growth factor receptor, alpha polypeptide
Kinases & Inhibitors
PDGFR Kinase Family
Human
GST Tag
Documents