PDGFRA [D842V] Recombinant Human Protein, Invitrogen™ DFS Item
PDGFRA is a cell surface receptor tyrosine kinase which binds members of the platelet-derived growth factor family.
Manufacturer:  Life Technologies LSPV4292
 View more versions of this product
Catalog No. PV4292
Description
Description
PDGFRA D842V is expressed in a high percentage of epithelial ovarian cancers and is implicated in the pathogenesis of acute myelogenous leukemia.
Specifications
Specifications
NP_006197, D842V
Frozen
89.7kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
PDGFR Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol
1mg
RUO - research use only
PDGFRA D842V, PDGFR alpha
Baculovirus
Human
GST Tag
Documents