Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ PDGFRα Ab-1, Rabbit Polyclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ PDGFRα Ab-1, Rabbit Polyclonal Antibody.

$272.78 - $1,269.59

Specifications

Antigen PDGFR, alpha Ab-1
Concentration 1mg/mL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB1691P

 
thermo scientific™ lab vision™
RB1691P
Purified Ab with BSA and Azide 1mL Each for $1,205.61

RB1691P0

 
thermo scientific™ lab vision™
RB1691P0
Purified Ab with BSA and Azide 100μL Each for $272.78

RB1691P1

 
thermo scientific™ lab vision™
RB1691P1
Purified Ab with BSA and Azide 500μL Each for $723.73

RB1691P1ABX

 
thermo scientific™ lab vision™
RB1691P1ABX
Ab without BSA/Azide (Frozen) 500μL Each for $806.08

RB1691PABX

 
thermo scientific™ lab vision™
RB1691PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,269.59
Description & Specifications

Specifications

Antigen PDGFR, alpha Ab-1
Concentration 1mg/mL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline Growth Factor
Type Primary
Immunogen A synthetic peptide derived from the C-terminus of the precursor form of human PDGFR, alpha
Isotype IgG

Provide accurate, reproducible results with the Thermo Scientific™ PDGFRα Ab-1, Rabbit Polyclonal Antibody.

PDGFR (Platelet Derived Growth Factor Receptor) exhibits two different isoforms, α and ß coded by two different genes. They possess five immunoglobulin like domains that are involved in ligand binding. Two tyrosine kinase domains are separated by a kinase insert to which PI-3 kinase can bind. PDGF AA can bind autophosphorylate and activate only PDGFR, alpha while PDGF BB can bind and activate to both PDGFR, α and β

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of the precursor form of human PDGFRα

Molecular Weight: ∽190kDa

Postitive Control: Ovarian CA

Cellular Localization: Membrane Specific for PDGFR, α and does not react with PDGFR, β.

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO