Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ PDGFRβ Ab-1, Rabbit Polyclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ PDGFRβ Ab-1, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1692PABX

 View more versions of this product

Catalog No. RB1692PABX

  • / Each
 

Description & Specifications

Specifications

Antigen PDGFR, beta Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Rabbit
Immunogen A synthetic peptide derived from near the C-terminus of human PDGFR, beta
Isotype IgG
Quantity 1mL
Regulatory Status RUO
Research Discipline Growth Factor
Type Primary
Monoclonal or Polyclonal Polyclonal
Target Species Human

Provide accurate, reproducible results with the Thermo Scientific™ PDGFRβ Ab-1, Rabbit Polyclonal Antibody.

PDGFR (Platelet Derived Growth Factor Receptor) exhibits two different isoforms, α and β coded by two different genes. They possess five immunoglobulin like domains that are involved in ligand binding. Two tyrosine kinase domains are separated by a kinase insert to which PI-3 kinase can bind. PDGF AA can bind autophosphorylate and activate only PDGFR, α while PDGF BB can bind and activate to both PDGFR, α and β.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not known.

Epitope: Near C-terminal

Immunogen: A synthetic peptide derived from near the C-terminus of human PDGFRβ

Molecular Weight: ∽190kDa

Postitive Control: Ovarian CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO