Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ PDGFRβ Ab-1, Rabbit Polyclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ PDGFRβ Ab-1, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1692P1ABX

 View more versions of this product

Catalog No. RB1692P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Paraffin)
Host Species Rabbit
Regulatory Status RUO
Quantity 500µL
Immunogen A synthetic peptide derived from near the C-terminus of human PDGFR, beta
Format Concentrated
Isotype IgG
Cross Reactivity Human
Antigen PDGFR, beta Ab-1
Classification Polyclonal
Formulation Ab without BSA/Azide (Frozen)
Conjugate Unlabeled
Research Discipline Growth Factor
Type Primary
Concentration 1mg/mL
Item Description 500uL; 1mg/mL; Unlabeled; Purified without BSA and azide

Provide accurate, reproducible results with the Thermo Scientific™ PDGFRβ Ab-1, Rabbit Polyclonal Antibody.

PDGFR (Platelet Derived Growth Factor Receptor) exhibits two different isoforms, α and β coded by two different genes. They possess five immunoglobulin like domains that are involved in ligand binding. Two tyrosine kinase domains are separated by a kinase insert to which PI-3 kinase can bind. PDGF AA can bind autophosphorylate and activate only PDGFR, α while PDGF BB can bind and activate to both PDGFR, α and β.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not known.

Epitope: Near C-terminal

Immunogen: A synthetic peptide derived from near the C-terminus of human PDGFRβ

Molecular Weight: ∽190kDa

Postitive Control: Ovarian CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO