missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ PDGFRβ Ab-1, Rabbit Polyclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ PDGFRβ Ab-1, Rabbit Polyclonal Antibody.

$276.86 - $1,311.59

Specifications

Concentration 1mg/mL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Format Concentrated
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
RB1692P Thermo Scientific™ Lab Vision™
RB1692P
Purified Ab with BSA and Azide 1mL Each for $1,217.59
RB1692P0 Thermo Scientific™ Lab Vision™
RB1692P0
Purified Ab with BSA and Azide 100μL Each for $276.86
RB1692P1 Thermo Scientific™ Lab Vision™
RB1692P1
Purified Ab with BSA and Azide 500μL Each for $735.69
RB1692P1ABX Thermo Scientific™ Lab Vision™
RB1692P1ABX
Ab without BSA/Azide (Frozen) 500μL Each for $818.14
RB1692PABX Thermo Scientific™ Lab Vision™
RB1692PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,311.59
Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ PDGFRβ Ab-1, Rabbit Polyclonal Antibody.

PDGFR (Platelet Derived Growth Factor Receptor) exhibits two different isoforms, α and β coded by two different genes. They possess five immunoglobulin like domains that are involved in ligand binding. Two tyrosine kinase domains are separated by a kinase insert to which PI-3 kinase can bind. PDGF AA can bind autophosphorylate and activate only PDGFR, α while PDGF BB can bind and activate to both PDGFR, α and β.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not known.

Epitope: Near C-terminal

Immunogen: A synthetic peptide derived from near the C-terminus of human PDGFRβ

Molecular Weight: ∽190kDa

Postitive Control: Ovarian CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

1mg/mL
Polyclonal
Concentrated
RUO
Primary
IgG
Immunohistochemistry (Paraffin)
Unlabeled
Rabbit
Growth Factor
A synthetic peptide derived from near the C-terminus of human PDGFR, beta