missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PD 407824, Tocris

Selective inhibitor of Chk1 and Wee1

$133.02 - $282.76

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C20H12N2O3
Formula Weight 328.32
Synonym 9-Hydroxy-4-phenyl-pyrrolo[3,4-c]carbazole-1,3(2H,6H)-dione
Chemical Name or Material PD 407824
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
26941 R & D Systems
26941
1mg Each for $133.02
269410 R & D Systems
269410
10mg Each for $282.76
Specifications Description & Specifications

Chemical Identifiers

>98%
328.32
PD 407824
C20H12N2O3
9-Hydroxy-4-phenyl-pyrrolo[3,4-c]carbazole-1,3(2H,6H)-dione

Specifications

1mg
>98%
328.32
PD 407824
622864-54-4
C20H12N2O3
9-Hydroxy-4-phenyl-pyrrolo[3,4-c]carbazole-1,3(2H,6H)-dione

Safety and Handling

Recommended Storage : Store at +4°C