Please login to your online account to display your discounted pricing

PD-1H Armenian Hamster anti-Mouse, PerCP-eFluor 710, Clone: MH5A, eBioscience™

Armenian Hamster Monoclonal Antibody

$201.55 - $491.55

Specifications

Antigen PD-1H
Clone MH5A
Host Species Armenian Hamster
Gene Alias VISTA
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123872

 
life technologies
LS46591980
25μg Each for $201.55

501123873

 
life technologies
LS46591982
100μg Each for $491.55
Description & Specifications

Specifications

Antigen PD-1H
Clone MH5A
Host Species Armenian Hamster
Gene Alias VISTA
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody MH5A reacts with a homolog of PD-1 (PD-1H) also known as VISTA. PD-1H is a 309 aa type I transmembrane protein that belongs to the CD28 superfamily. This cell surface receptor is expressed on nearly all hematopoietic cells except B cells. VISTA negatively regulates T cell responses by suppressing early TCR activation, inhibiting cell proliferation, and suppressing TH1 cytokine production.

The MH5A antibody has been reported to inhibit T cell infiltration in graft-versus-host disease mouse models.