Please login to your online account to display your discounted pricing

PD 184352, Tocris™

Selective MEK inhibitor

$303.99 - $1,257.94

Chemical Identifiers

Chemical Name or Material PD 184352
Assay Percent Range >98%
Molecular Formula C17H14ClF2IN2O2
Formula Weight 478.66
Synonym 2-[(2-Chloro-4-iodophenyl)amino]-N-cyclopropylmethoxy)-3, 4-difluorobenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

423710

 
r & d systems
RDS423710
10mg Each for $303.99

423750

 
r & d systems
RDS423750
50mg Each for $1,257.94
Description & Specifications

Specifications

Formula Weight 478.66
Quantity 10mg
Chemical Name or Material PD 184352
Assay Percent Range >98%
Molecular Formula C17H14ClF2IN2O2
CAS 212631-79-3
Synonym 2-[(2-Chloro-4-iodophenyl)amino]-N-cyclopropylmethoxy)-3, 4-difluorobenzamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C