Please login to your online account to display your discounted pricing

PD 173074, Tocris

FGFR1 and -3 inhibitor

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material PD 173074
Assay Percent Range >98%
Molecular Formula C28H41N7O3
Formula Weight 523.67
Synonym N-[2-[[4-(Diethylamino)butyl]amino]-6-(3,5-dimethoxyphenyl)pyrido[2,3-d]pyrimidin-7-yl]-N'-(1,1-dimethylethyl)urea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

304410

 
r & d systems
304410
10mg Each for $252.39

304450

 
r & d systems
304450
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 523.67
Quantity 10mg
Assay Percent Range >98%
Molecular Formula C28H41N7O3
Synonym N-[2-[[4-(Diethylamino)butyl]amino]-6-(3,5-dimethoxyphenyl)pyrido[2,3-d]pyrimidin-7-yl]-N'-(1,1-dimethylethyl)urea
Chemical Name or Material PD 173074
CAS 219580-11-7
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C