Please login to your online account to display your discounted pricing

PD 168077 maleate, Tocris


High affinity, selective D4 agonist

Quantity 10mg

Manufacturer: r & d systems  106510

 View more versions of this product

Catalog No. 106510

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 450.49
Quantity 10mg
CAS 630117-19-0
Synonym N-(Methyl-4-(2-cyanophenyl)piperazinyl-3-methylbenzamide maleate
Chemical Name or Material PD 168077 maleate
Assay Percent Range >99%
Molecular Formula C20H22N4O·C4H4O4
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material PD 168077 maleate
Assay Percent Range >99%
Molecular Formula C20H22N4O·C4H4O4
Formula Weight 450.49
Synonym N-(Methyl-4-(2-cyanophenyl)piperazinyl-3-methylbenzamide maleate
Accessories

Desiccate at -20°C