missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PD 166285 dihydrochloride, Tocris™

Potent Src inhibitor; also inhibits FGFR1, PDGFRβ

$133.02 - $282.76

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C26H27Cl2N5O2·2HCl
Formula Weight 585.35
Synonym 6-(2, 6-Dichlorophenyl)-2-[[4-[2-(diethylamino)ethoxy]phenyl]amino]-8-methylpyrido[2, 3-d]pyrimidin-7(8H)-one dihydrochloride
Chemical Name or Material PD 166285 dihydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
37851 R & D Systems
RDS37851
1mg Each for $133.02
378510 R & D Systems
RDS378510
10mg Each for $282.76
Specifications Description & Specifications

Chemical Identifiers

>99%
585.35
PD 166285 dihydrochloride
C26H27Cl2N5O2·2HCl
6-(2, 6-Dichlorophenyl)-2-[[4-[2-(diethylamino)ethoxy]phenyl]amino]-8-methylpyrido[2, 3-d]pyrimidin-7(8H)-one dihydrochloride

Specifications

1mg
>99%
585.35
PD 166285 dihydrochloride
212391-63-4
C26H27Cl2N5O2·2HCl
6-(2, 6-Dichlorophenyl)-2-[[4-[2-(diethylamino)ethoxy]phenyl]amino]-8-methylpyrido[2, 3-d]pyrimidin-7(8H)-one dihydrochloride

Safety and Handling

Recommended Storage : Desiccate at Room Temperature