missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PD 160170, Tocris

Selective non-peptide NPY Y1 receptor antagonist

$212.57 - $889.32

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C18H17N3O4S
Formula Weight 371.41
Synonym 6-[[2-(1-Methylethyl)phenyl]]sulfonyl-5-nitro-8-quinolinamine
Chemical Name or Material PD 160170
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
220050 R & D Systems
RDS220050
50mg Each for $889.32
220010 R & D Systems
RDS220010
10mg Each for $212.57
Specifications Description & Specifications

Chemical Identifiers

>98%
371.41
PD 160170
C18H17N3O4S
6-[[2-(1-Methylethyl)phenyl]]sulfonyl-5-nitro-8-quinolinamine

Specifications

50mg
>98%
371.41
PD 160170
181468-88-2
C18H17N3O4S
6-[[2-(1-Methylethyl)phenyl]]sulfonyl-5-nitro-8-quinolinamine

Safety and Handling

Recommended Storage : Store at Room Temperature