Please login to your online account to display your discounted pricing

PD 158780, Tocris

Potent ErbB receptor family inhibitor

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material PD 158780
Assay Percent Range >99%
Molecular Formula C14H12BrN5
Formula Weight 330.18
Synonym N4-(3-Bromophenyl)-N6-methyl-pyrido[3, 4-d]pyrimidine-4, 6-diamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

261510

 
r & d systems
261510
10mg Each for $239.63

261550

 
r & d systems
261550
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 330.18
Quantity 10mg
Chemical Name or Material PD 158780
Assay Percent Range >99%
Molecular Formula C14H12BrN5
CAS 171179-06-9
Synonym N4-(3-Bromophenyl)-N6-methyl-pyrido[3, 4-d]pyrimidine-4, 6-diamine
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C