Please login to your online account to display your discounted pricing

PD 150606, Tocris


Cell permeable calpain inhibitor

Quantity 10mg

Manufacturer: r & d systems  RDS126910

 View more versions of this product

Catalog No. 126910

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 306.12
Quantity 10mg
Chemical Name or Material PD 150606
Assay Percent Range >95%
Molecular Formula C9H7IO2S
CAS 179528-45-1
Synonym (Z)-3-(4-Iodophenyl)-2-mercapto-2-propenoic acid
Recommended Storage Store at -20°C

Specifications

Chemical Name or Material PD 150606
Assay Percent Range >95%
Molecular Formula C9H7IO2S
Formula Weight 306.12
Synonym (Z)-3-(4-Iodophenyl)-2-mercapto-2-propenoic acid
Accessories

Store at -20°C