Please login to your online account to display your discounted pricing

PD 150606, Tocris

Cell permeable calpain inhibitor

$199.70 - $834.54

Chemical Identifiers

Chemical Name or Material PD 150606
Assay Percent Range >95%
Molecular Formula C9H7IO2S
Formula Weight 306.12
Synonym (Z)-3-(4-Iodophenyl)-2-mercapto-2-propenoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

126910

 
r & d systems
RDS126910
10mg Each for $199.70

126950

 
r & d systems
RDS126950
50mg Each for $834.54
Description & Specifications

Specifications

Formula Weight 306.12
Quantity 10mg
Chemical Name or Material PD 150606
Assay Percent Range >95%
Molecular Formula C9H7IO2S
CAS 179528-45-1
Synonym (Z)-3-(4-Iodophenyl)-2-mercapto-2-propenoic acid
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C