Please login to your online account to display your discounted pricing

PD 135158, Tocris

Potent and selective CCK2 antagonist

$323.08 - $1,333.44

Chemical Identifiers

Chemical Name or Material PD 135158
Assay Percent Range >98%
Molecular Formula C35H44N4O6
Formula Weight 616.76
Synonym 4-[[(1R)-2-[[(2R)-3-(1H-Indol-3-yl)-2-methyl-1-oxo-2-[[[[(1S,2R,4S)-1,7,7-trimethylbicyclo[2.2.1]hept-2-yl]oxy]carbonyl]amino]propyl]amino]-1-phenylethyl]amino]-4-oxobutanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

260810

 
r & d systems
260810
10mg Each for $323.08

260850

 
r & d systems
260850
50mg Each for $1,333.44
Description & Specifications

Specifications

Formula Weight 616.76
Quantity 10mg
Molecular Formula C35H44N4O6
Assay Percent Range >98%
Chemical Name or Material PD 135158
CAS 130285-87-9
Synonym 4-[[(1R)-2-[[(2R)-3-(1H-Indol-3-yl)-2-methyl-1-oxo-2-[[[[(1S,2R,4S)-1,7,7-trimethylbicyclo[2.2.1]hept-2-yl]oxy]carbonyl]amino]propyl]amino]-1-phenylethyl]amino]-4-oxobutanoic acid
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C