Please login to your online account to display your discounted pricing

PD 123319 ditrifluoroacetate, Tocris

Potent, selective non-peptide AT2 antagonist

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material PD 123319 ditrifluoroacetate
Assay Percent Range >99%
Molecular Formula C31H32N4O3·2CF3CO2H
Formula Weight 736.67
Synonym 1-[[4-(Dimethylamino)-3-methylphenyl]methyl]-5-(diphenylacetyl)-4,5,6,7-tetrahydro-1H-imidazo[4,5-c]pyridine-6-carboxylic acid ditrifluoroacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

136110

 
r & d systems
136110
10mg Each for $257.85

136150

 
r & d systems
136150
50mg Each for $1,073.10
Description & Specifications

Specifications

Formula Weight 736.67
Quantity 10mg
Chemical Name or Material PD 123319 ditrifluoroacetate
CAS 130663-39-7
Assay Percent Range >99%
Molecular Formula C31H32N4O3·2CF3CO2H
Synonym 1-[[4-(Dimethylamino)-3-methylphenyl]methyl]-5-(diphenylacetyl)-4,5,6,7-tetrahydro-1H-imidazo[4,5-c]pyridine-6-carboxylic acid ditrifluoroacetate
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C