Please login to your online account to display your discounted pricing
Thermo Scientific™ PCR Plate, 96-Well, Semi-Skirted, Flat Deck, Black Lettering
Thermo Scientific™ PCR plastics are designed, manufactured, and tested to ensure PCR performance
$240.18 - $240.18
Products
Catalog Number Mfr. No. Color Price Quantity    
AB1400BL
thermo scientific™
AB1400BL
Blue Case for $240.18
AB1400GL
thermo scientific™
AB1400GL
Green Case for $240.18
AB1400PL
thermo scientific™
AB1400PL
Purple Case for $240.18
AB1400RL
thermo scientific™
AB1400RL
Red Case for $240.18
AB1400YL
thermo scientific™
AB1400YL
Yellow Case for $240.18
Description & Specifications
The Thermo Scientific™ 96-Well, Black Lettering, Semi-Skirted Flat Deck PCR Plates are compatible with most thermal cyclers and ABI platforms, including sequencers, with no adapters necessary. The Thermo Scientific PCR plastics are designed, manufactured, and tested to ensure PCR performance. The plates have 0.3 mL maximum well volume when used with adhesive and heat seals
Features of 96-Well Semi-Skirted Flat Deck PCR Plates
  • Directly compatible with all major major thermal cyclers, including ABI platforms and sequencers with no adapters necessary
  • Black lettering facilitates sample tracking
  • Flat plate deck facilitates sealing and handling
  • Available as SuperPlate providing 4X more rigidity for superior robotic handling
  • Raised rim design around each well enables secure sealing and safeguards against evaporation
  • Uniform, thin well walls deliver maximum and consistent heat transfer
  • Black lettered versions feature black alpha-numeric lettering to allow for easy well identification
  • White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
  • Cut corner: A12
Applications
  • PCR