Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ PCNA (Proliferating Cell Nuclear Antigen) Ab-1, Mouse Monoclonal Antibody

Provides accurate, reproducible results with the Thermo Scientific™ PCNA (Proliferating Cell Nuclear Antigen) Ab-1, Mouse Monoclonal Antibody.

$277.84 - $1,325.33

Specifications

Antigen PCNA (Proliferating Cell Nuclear Antigen) Ab-1
Clone PC10
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS106B

 
thermo scientific™ lab vision™
MS106B
Biotinylated Ab with BSA and Azide 1mL Each for $1,325.33

MS106B0

 
thermo scientific™ lab vision™
MS106B0
Biotinylated Ab with BSA and Azide 100µL Each for $378.20

MS106B1

 
thermo scientific™ lab vision™
MS106B1
Biotinylated Ab with BSA and Azide 500µL Each for $898.53

MS106P

 
thermo scientific™ lab vision™
MS106P
Purified Ab with BSA and Azide 1mL Each for $1,031.14

MS106P0

 
thermo scientific™ lab vision™
MS106P0
Purified Ab with BSA and Azide 100µL Each for $277.84

MS106P1

 
thermo scientific™ lab vision™
MS106P1
Purified Ab with BSA and Azide 500µL Each for $683.31

MS106P1ABX

 
thermo scientific™ lab vision™
MS106P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $774.06

MS106PABX

 
thermo scientific™ lab vision™
MS106PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,127.16
Description & Specifications

Specifications

Antigen PCNA (Proliferating Cell Nuclear Antigen) Ab-1
Clone PC10
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Cross Reactivity Human
Cross Reactivity Insect
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Yeast
Cross Reactivity Human
Cross Reactivity Insect
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Yeast
Cross Reactivity Human
Cross Reactivity Insect
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Yeast
Cross Reactivity Human
Cross Reactivity Insect
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Yeast
Cross Reactivity Human
Cross Reactivity Insect
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Yeast
Host Species Mouse
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Immunogen Recombinant rat PCNA protein
Isotype IgG2a kappa

Provides accurate, reproducible results with the Thermo Scientific™ PCNA (Proliferating Cell Nuclear Antigen) Ab-1, Mouse Monoclonal Antibody.

Expression of proliferating cell nuclear antigen (PCNA) or cyclin or polymerase δ auxiliary protein is elevated in the nucleus during late G1 phase immediately before the onset of DNA synthesis, becoming maximal during S-phase and declining during G2 and M phases. Its level correlates directly with rates of cellular proliferation and DNA synthesis. PCNA/cyclin may act as an auxiliary protein of DNA polymerase-δ to play a fundamental role in the initiation of cell proliferation

Host Species: Mouse

Clone: PC10

Isotype: IgG2a/κ

Species Reactivity: Human, Mouse, Rat, Insect, and Yeast. Others not known.

Epitope: Not determined

Immunogen: Recombinant rat PCNA protein

Molecular Weight: 36kDa

Positive Control: Raji cells; Tonsil, lymph node, or small intestine

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO