Learn More
Epredia™ Lab Vision™ PCNA (Proliferating Cell Nuclear Antigen) Ab-1 Mouse Monoclonal Antibody, 20μg/mL, biotinylated, BSA and azide
Provides accurate, reproducible results with the Epredia™ PCNA (Proliferating Cell Nuclear Antigen) Ab-1, Mouse Monoclonal Antibody.
Supplier: Epredia™ MS106P1ABX
Description
Expression of proliferating cell nuclear antigen (PCNA) or cyclin or polymerase δ auxiliary protein is elevated in the nucleus during late G1 phase immediately before the onset of DNA synthesis, becoming maximal during S-phase and declining during G2 and M phases. Its level correlates directly with rates of cellular proliferation and DNA synthesis. PCNA/cyclin may act as an auxiliary protein of DNA polymerase-δ to play a fundamental role in the initiation of cell proliferationHost Species: Mouse
Clone: PC10
Isotype: IgG2a/κ
Species Reactivity: Human, Mouse, Rat, Insect, and Yeast. Others not known.
Epitope: Not determined
Immunogen: Recombinant rat PCNA protein
Molecular Weight: 36kDa
Positive Control: Raji cells; Tonsil, lymph node, or small intestine
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native and denatured)
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
PCNA (Proliferating Cell Nuclear Antigen) Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Cell Cycle | |
Human, Insect, Mouse, Rat, Yeast | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
PC10 | |
Unconjugated | |
Recombinant rat PCNA protein | |
RUO | |
Primary | |
Concentrated |