missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PCNA Mouse anti-Human, Clone: PC10 (a.k.a. 3F81), eBioscience™

Mouse Monoclonal Antibody

$85.55 - $187.05

Specifications

Antigen PCNA
Clone PC10 (a.k.a. 3F81)
Host Species Murine
Gene Alias Proliferating Cell Nuclear Antigen
Species Reactivity Human, Mouse, Rat
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013850 Life Technologies
LS14991080
25μg Each for $85.55
5013851 Life Technologies
LS14991082
100μg Each for $187.05
Specifications

Description

The PC10 antibody recognizes the proliferating cell nuclear antigen (PCNA), a 36kDa protein, also known as polymerase delta auxiliary protein. PC10 antibody reacts with human, mouse, and rat PCNA. The peak expression of PCNA occurs during the S-phase.
Description & Specifications

Specifications

PCNA
Murine
Human, Mouse, Rat
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
PC10 (a.k.a. 3F81)
Proliferating Cell Nuclear Antigen
Flow Cytometry,Immunocytochemistry,Immunohistochemistry,Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL