Please login to your online account to display your discounted pricing

PCA 4248, Tocris


PAF receptor antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS057110

 View more versions of this product

Catalog No. 057110

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 361.46
Chemical Name or Material PCA 4248
Molecular Formula C19H23NO4S
Assay Percent Range >95%
CAS 123875-01-4
Synonym 1,4-Dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylic acid methyl 2-(phenylthio)ethyl ester
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material PCA 4248
Assay Percent Range >95%
Molecular Formula C19H23NO4S
Formula Weight 361.46
Synonym 1,4-Dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylic acid methyl 2-(phenylthio)ethyl ester
Accessories

Desiccate at -20°C