Please login to your online account to display your discounted pricing

PCA 4248, Tocris

PAF receptor antagonist

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material PCA 4248
Assay Percent Range >95%
Molecular Formula C19H23NO4S
Formula Weight 361.46
Synonym 1,4-Dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylic acid methyl 2-(phenylthio)ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

057110

 
r & d systems
RDS057110
10mg Each for $164.61

057150

 
r & d systems
RDS057150
50mg Each for $682.67
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 361.46
Chemical Name or Material PCA 4248
Assay Percent Range >95%
Molecular Formula C19H23NO4S
CAS 123875-01-4
Synonym 1,4-Dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylic acid methyl 2-(phenylthio)ethyl ester
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C